Order Entry
Italy
ContactUsLinkComponent
Indo-1 pentammonium salt ≥95% (by TLC) fluorescent indicator, for calcium determination
Indo-1 pentammonium salt ≥95% (by TLC) fluorescent indicator, for calcium determination
Catalog # 50040.
Supplier:  Biotium
CAS Number:  
Indo-1 pentammonium salt ≥95% (by TLC) fluorescent indicator, for calcium determination
Catalog # 50040.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :735 g/mol
Storage Temperature:Refrigerator
 :132319-56-3
Formula:C₃₂H₄₆N₈O₁₂