Rhod-590 AM ester fluorescent indicator, for calcium determination
Catalog # 50025.
Supplier: Biotium
CAS Number:
Specifications
- Pk:500 µG
- Seq:1418
- Environmentally Preferable:
- Pk Info:10 x 50 µG
Specifications
About this item
:
:1204 g/mol
Storage Temperature:Freezer
Formula:C₆₄H₇₅N₄O₁₉