5,5’-Dimethyl BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Codice catalogo 50008.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:657 g/mol
Storage Temperature:Freezer
CAS Number:73630-09-8
Formula:C₂₄H₂₄K₄N₂O₁₀