Order Entry
Italy
ContactUsLinkComponent
5,5’-Dimethyl BAPTA AM ester ≥95% (by TLC) ion chelator, for calcium determination
5,5’-Dimethyl BAPTA AM ester ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50007.
Supplier:  Biotium
CAS Number:  
5,5’-Dimethyl BAPTA AM ester ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50007.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :792.75 g/mol
Storage Temperature:Freezer
CAS Number:147504-94-7
Formula:C₃₆H₄₄N₂O₁₈