Order Entry
Italy
ContactUsLinkComponent
5,5′-Dibromo BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
5,5′-Dibromo BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50004.
Supplier:  Biotium
CAS Number:  
5,5′-Dibromo BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50004.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :786.59 g/mol
Storage Temperature:Refrigerator
CAS Number:73630-11-2
Formula:C₂₂H₁₈Br₂K₄N₂O₁₀
MDL Number:MFCD00036169