Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Demand-WP, (CRM), Supelco®
Demand-WP, (CRM), Supelco®
Catalog # QC1130-20ML
Supplier:
Merck
CAS Number:
Certificates
Expand image
Demand-WP, (CRM), Supelco®
Catalog # QC1130-20ML
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
20 mL
Description:
Demand-WP
Type:
Certified reference material
Specifications
About this item
Product Details
Documentation
Product Details
Documentation