Supelco® Organic Reference Standard, MDI-DBA Oligomers Standard
Catalog # CRM40603
Supplier: Merck
CAS Number:
Specifications
- Concentration:In methanol (varied conc.)
- Pk:1 mL
- Standards:Certified reference material
- Description:MDI-DBA oligomers standard
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):Methylenediphenyl diisocyanate dibutylamine oligomers standard; p-MDI; polymeric-MDI
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311