Order Entry
Italy
ContactUsLinkComponent
ISO 17943 57 Component VOC mix, (CRM), Supelco®
ISO 17943 57 Component VOC mix, (CRM), Supelco®
Catalog # 44926-U
Supplier:  Merck
CAS Number:  
ISO 17943 57 Component VOC mix, (CRM), Supelco®
Catalog # 44926-U
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    200 µg/ml each component in methanol
  • Pk:
    1 Ampoul
  • Description:
    ISO 17943 57 Component VOC Mix
  • Type:
    Certified reference material

Specifications

About this item