Order Entry
Italy
ContactUsLinkComponent
5-CFDA (5-Carboxyfluorescein diacetate) ≥99% (by HPLC) fluorescent pH indicator
5-CFDA (5-Carboxyfluorescein diacetate) ≥99% (by HPLC) fluorescent pH indicator
Catalog # 51018.
Supplier:  Biotium
CAS Number:  
undefined
5-CFDA (5-Carboxyfluorescein diacetate) ≥99% (by HPLC) fluorescent pH indicator
Catalog # 51018.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :460.39 g/mol
Storage Temperature:Refrigerator
CAS Number:79955-27-4
Formula:C₂₅H₁₆O₉
MDL Number:MFCD00036872