Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Dextromethorphan Impurity C
Supelco® Organic Reference Standard, Dextromethorphan Impurity C
Catalog # PHR1975-30MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Dextromethorphan Impurity C
Catalog # PHR1975-30MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    30 mg
  • Standards:
    Pharmaceutical Secondary Standard; Certified Reference Material
  • CAS No.:
    57969-05-8
  • Description:
    Dextromethorphan impurity C
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 - H336
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (9α,13α,14α)-3-Methoxy-17-methylmorphinan-10-one; 10-Keto dextromethorphan
  • Molecular weight:
    285,38

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P271 - P301 + P310 - P304 + P340 + P312