0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Paclobutrazol, (CRM), TraceCERT®, Supelco®
Organic reference standard, Paclobutrazol, (CRM), TraceCERT®, Supelco®
43900-50MG
:
Merck
:
Organic reference standard, Paclobutrazol, (CRM), TraceCERT®, Supelco®
43900-50MG
:
Merck
:
Concentration:
Neat
Pk:
50 mg
CAS No.:
76738-62-0
Description:
Paclobutrazol
Description:
Paclobutrazol