Order Entry
Italy
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, 18-Hydroxycorticosterone solution
Supelco® Cerilliant® Organic Reference Standard, 18-Hydroxycorticosterone solution
Catalog # H-106-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, 18-Hydroxycorticosterone solution
Catalog # H-106-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile: water (9:1)
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    561-65-9
  • Description:
    18-Hydroxycorticosterone solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00042685
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    362,46
  • Flash point:
    2,0 °C - Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338