Order Entry
Italy
ContactUsLinkComponent
Pharmaceutical secondary standard, Benzalkonium Chloride 50% Solution, (CRM), Supelco®
Pharmaceutical secondary standard, Benzalkonium Chloride 50% Solution, (CRM), Supelco®
Catalog # PHR1681-1ML
Supplier:  Merck
CAS Number:  
undefined
Pharmaceutical secondary standard, Benzalkonium Chloride 50% Solution, (CRM), Supelco®
Catalog # PHR1681-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Benzalkonium Chloride 50% Solution

Specifications

About this item