Order Entry
Italy
ContactUsLinkComponent
 
Pharmaceutical secondary standard, Benzalkonium Chloride 10% Solution, (CRM), Supelco®
Catalog # PHR1371-5ML
Supplier:  Merck
CAS Number:  
Pharmaceutical secondary standard, Benzalkonium Chloride 10% Solution, (CRM), Supelco®
Catalog # PHR1371-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Description:
    Benzalkonium Chloride 10% Solution

Specifications

About this item