Order Entry
Italy
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, 2,4-D
Supelco® TraceCERT® Organic Reference Standard, 2,4-D
  76514-100MG
 :  Merck
 :  
undefined
Supelco® TraceCERT® Organic Reference Standard, 2,4-D
  76514-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    94-75-7
  • Description:
    2,4-D
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302 - H317 - H318 - H335 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004300
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    2,4-Dichlorophenoxyacetic acid; 2,4-D
  • Melting point:
    136 - 140 °C (lit.)
  • Formula:
    Cl₂C₆H₃OCH₂CO₂H
  • Molecular weight:
    221,04

 

 

  • GHS signal: GHS05, GHS07
 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P261 - P273 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338