Inserisci ordine
Italy
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Carbamazepine-10,11-epoxide-13C6 solution, (CRM)
Supelco® Drug and metabolite standards, Carbamazepine-10,11-epoxide-13C6 solution, (CRM)
Catalog # C-128-1ML
Supplier:  Merck
CAS Number:  
Supelco® Drug and metabolite standards, Carbamazepine-10,11-epoxide-13C6 solution, (CRM)
Catalog # C-128-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentrazione:
    100 µg/ml in methanol
  • Conf:
    1 mL
  • Standard:
    Certified reference material
  • Descrizione:
    Carbamazepine-10
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item