Order Entry
Italy
ContactUsLinkComponent
Drug and metabolite standards, 6beta-Hydroxytestosterone solution, (CRM), Supelco®
Drug and metabolite standards, 6beta-Hydroxytestosterone solution, (CRM), Supelco®
Catalog # H-059-1ML
Supplier:  Merck
CAS Number:  
undefined
Drug and metabolite standards, 6beta-Hydroxytestosterone solution, (CRM), Supelco®
Catalog # H-059-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    62-99-7
  • Description:
    6beta-Hydroxytestosterone solution
  • Description:
    6beta-Hydroxytestosterone solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item