Order Entry
Italy
ContactUsLinkComponent
 
Supelco® Cerilliant® Organic Reference Standard, 11-Deoxycortisol solution
Catalog # D-061-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, 11-Deoxycortisol solution
Catalog # D-061-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    152-58-9
  • Description:
    11-Deoxycortisol solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00003662
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    346,46
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P260 - P280 - P301 + P310 - P311