Order Entry
Italy
ContactUsLinkComponent
Gel filtration chromatography columns, HiScale™ Superdex™ Increase and Superose™ Increase
Gel filtration chromatography columns, HiScale™ Superdex™ Increase and Superose™ Increase
Supplier:  Cytiva
Certificates
undefined
Gel filtration chromatography columns, HiScale™ Superdex™ Increase and Superose™ Increase
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 29-3219-04
  • 29-3219-03
  • 29-3219-06
  • 29-3219-05
  • 29-3219-08
  • 29-3219-07
  • Max. pressure:
  • 20 bar
  • 20 bar
  • 20 bar
  • 20 bar
  • 20 bar
  • 20 bar
  • Storage:
  • 4 - 30 °C, 20% ethanol
  • 4 - 30 °C, 20% ethanol
  • 4 - 30 °C, 20% ethanol
  • 4 - 30 °C, 20% ethanol
  • 4 - 30 °C, 20% ethanol
  • 4 - 30 °C, 20% ethanol