Columns, HiTrap™ Capto PlasmidSelect
About this item
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- Bioprocess resin supported for industrial applications
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.
2 Options Available Below
Product Details & Documents
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- Bioprocess resin supported for industrial applications
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications for Product Family
Specifications
- Catalog No:
- 17-5499-11
- 17-5499-12
- Max. pressure:
- 5 bar
- 5 bar
Specifications
Product Family Options
Product Information
- Bed dimensionsCapacityFlow rateAvailabilityPrice
- 17-5499-11HiTrap™ Capto PlasmidSelectSpecifications:
More
Specifications
Cat. No.17-5499-11Max. pressure5 bar - 17-5499-12HiTrap™ Capto PlasmidSelectSpecifications:
More
Specifications
Cat. No.17-5499-12Max. pressure5 bar