Order Entry
Italy
ContactUsLinkComponent
 
TLC plates, silica gel 60, precoated
  CAMA034.1798
 :  CAMAG
TLC plates, silica gel 60, precoated
  CAMA034.1798
 :  CAMAG

 

  • Coating material:
    Silica gel
  • Layer thickness:
    250 µm
  • Plate size:
    200×200 mm
  • Pore size:
    60 Å
  • Seq:
    01
  • Fluorescent indicator:
    F254
  • Concentrating zone:
    Yes
  • Pk:
    25

 

 

TLC precoated sheets, silica gel 60.