Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, N-nitroso-di-isopropylamine, Dr Ehrenstorfer
Organic reference standard, N-nitroso-di-isopropylamine, Dr Ehrenstorfer
Catalog # EHERC15604700
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, N-nitroso-di-isopropylamine, Dr Ehrenstorfer
Catalog # EHERC15604700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    601-77-4
  • Description:
    N-Nitroso-di-isopropylamine
  • Formula:
    C₆H₁₄N₂O
  • Description:
    N-Nitroso-di-isopropylamine

Specifications

About this item

Certifications: Produced in accordance to ISO 17034