Order Entry
Italy
ContactUsLinkComponent
Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis) USP, BP, Ph. Eur., pharma grade
Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates
Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (Compl.) (calc. A.A.S):
    99.0 - 100.5%
  • Identity: Identity according to Pharmacopoeias:
    Passes Test
  • pH of 5% Solution:
    5.0 - 9.2
  • Appearance of solution:
    Passes Test
  • Maximum Limit of Impurities, Acidity or Alkalinity:
    Passes Test
  • Maximum Limit of Impurities, Insoluble Matter In H₂O:
    0.025 %
  • Maximum Limit of Impurities, Loss on Drying at 450°C:
    48.0 - 52.0 %
  • Maximum Limit of Impurities, Chloride (Cl):
    0.014%
  • Maximum Limit of Impurities, Ammonium (NH₄):
    0.005%
  • Maximum Limit of Impurities, Phosphate (PO₄):
    0.002 %
  • Maximum Limit of Impurities, Residual Solvents (Ph. Eur./USP):
    passes test
  • Maximum Limit of Impurities, Heavy Metals (as Pb):
    0.001%
  • Maximum Limit of Impurities, Fe:
    0.002 %
  • Maximum Limit of Impurities, Se:
    0.0015 %
  • Maximum Limit of Impurities, As:
    0.00015 %
  • Elemental Impurities (ICH Q3D), Class 1, Cd:
    0.5 ppm
  • Elemental Impurities (ICH Q3D), Class 1, Pb:
    50 ppm
  • Elemental Impurities (ICH Q3D), Class 1, As:
    1.5 ppm
  • Elemental Impurities (ICH Q3D), Class 1, Hg:
    1.5 ppm
  • Elemental Impurities (ICH Q3D), Class 2A, Co:
    5 ppm
  • Elemental Impurities (ICH Q3D), Class 2A, V:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2A, Ni:
    20 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Au:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Pd:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Ir:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Os:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Rh:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Ru:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Se:
    15 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Ag:
    15 ppm
  • Elemental Impurities (ICH Q3D), Class 2B, Pt:
    10 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Li:
    55 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Sb:
    120 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Ba:
    140 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Mo:
    25 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Cu:
    250 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Sn:
    600 ppm
  • Elemental Impurities (ICH Q3D), Class 3, Cr:
    25 ppm

Specification Test Results

Frequently Bought Together