Sand (sea sand), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:14808-60-7
Formula:SiO₂
Boiling Pt:2230
Molecular Weight:60.08 g/mol
Melting Pt:1710
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Substances soluble in HCl:max 0.5%
- Chloride (Cl) :max. 0.01%
- Loss on ignition (900 °C):max. 0.1%
- Particle size (0,1 - 0,315 mm):min.90%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC86154.295 spec_VWRC86154.295 spec_VWRC86154.295 spec_VWRC86154.364 spec_VWRC86154.364
Product specification
More
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together