Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, polyhexamethylenbiguanid hydrochloride, Dr Ehrenstorfer
Organic reference standard, polyhexamethylenbiguanid hydrochloride, Dr Ehrenstorfer
Catalog # EHERC16282500
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, polyhexamethylenbiguanid hydrochloride, Dr Ehrenstorfer
Catalog # EHERC16282500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Standards:
    Reference standard
  • CAS No.:
    32289-58-0
  • Description:
    Polyhexamethylenbiguanid hydrochloride
  • Formula:
    (C₈H₁₇N₅)n . HCl
  • Description:
    Polyhexamethylenbiguanid hydrochloride

Specifications

About this item

Certifications: Accredited to ISO/IEC 17025.