Organic reference standard, doxycycline hyclate, Dr. Ehrenstorfer
Catalog # EHERC13084280
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,1 g
- Standards:Reference standard
- CAS No.:24390-14-5
- Description:Doxycycline hyclate
- Formula:2C₂₂H₂₄N₂O₈·C₂H₆O·2HCl·H₂O
- Description:Doxycycline hyclate
Specifications
About this item
Certifications: ISO/IEC 17025 certified.