
α-Naphtholphthalein indicator, Fluka™
33424-10G
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Change range (pH 7.1-8.3):complying
- Test of sensitivity:complying
:
:418.45 g/mol
:238-240 °C
:209-875-5
:GHS07
:Ambient
:596-01-0
:C₂₈H₁₈O₄
:MFCD00036202
:238…240 °C