
Aluminium sulphate octadecahydrate Al₂(SO₄)₃ 51.0-59.0% 100-110%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
:7784-31-8
:
:Al₂(SO₄)₃·18H₂O
:MFCD00149136
:666.43 g/mol
:87 °C
:1.7 g/cm3 (20 °C)
:233-135-0
:GHS05
:Ambient
- Assay:100.0 - 110.0 %
- Assay of Al₂(SO₄)3:51.0 - 59.0 %
- pH (2 %,20°C):2.5 - 4.0
- Ammonium (NH₄):Max. 30 ppm
- Arsenic (As):Max. 3 ppm
- Iron (Fe):Max. 50 ppm
- Heavy metals (as Pb):Max. 20 ppm
- Chloride (Cl):Max. 0.005 %
- Alkali+alkaline earth metals:Max. 0.2 %
- Appearance of the solution:complying
- Residual solvents:complying
Frequently Bought Together