Order Entry
Italy
ContactUsLinkComponent
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Irritant
Flammable
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Certificates
undefined
Irritant
Flammable
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™

Specification Test Results

  • factor:
    0.998 - 1.002
  • Identity (IR):
    complying
  • Identity (NMR):
    complying
  • Assay (GC):
    Min. 98.5 %
  • Density (D 20/4):
    0.723 - 0.725
  • Refractive index (n 20/D):
    1.367 - 1.369
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as C₂H₅COOH):
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • 2-propanol (GC):
    Max. 0.5 %
  • Assay BHT (GC):
    0.0005 - 0.02 %
  • Peroxides (as H₂O₂):
    Max. 10 ppm
  • APHA:
    Max. 25
  • Autoignition Temperature:
    827 °F

Specification Test Results

Related Information

Frequently Bought Together