Order Entry
Italy
ContactUsLinkComponent
Barium hydroxide octahydrate ≥98%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Corrosive
Irritant
Barium hydroxide octahydrate ≥98%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Certificates
Corrosive
Irritant
Barium hydroxide octahydrate ≥98%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™

Specification Test Results

  • Assay:
    Min. 98 %
  • Assay of BaCO₃:
    Max. 2 %
  • Insoluble in acid:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.002 %
  • Sodium (Na):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Strontium (Sr):
    Max. 1.5 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfide (S):
    Max. 0.0005 %

Specification Test Results

Related Information

Frequently Bought Together