Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
2-Ethoxyethanol 99%, Reagent Grade
 
undefined
 
 
 
2-Ethoxyethanol 99%, Reagent Grade

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 98.5 %
  • Water (Karl Fischer):
    Max. 0.10 %
  • APHA:
    Max. 10
  • Autoignition Temperature:
    460 °F
  • Refractive Index:
    n20/D 1.407 (lit.)
  • Vapor Pressure:
    3.8 mmHg ( 20 °C)

 

 

Frequently Bought Together