- Environmentally Preferable:
- Assay of active chlorine:4 - 5 %
- Vapor Pressure:17.5 mmHg ( 20 °C)
- Composition:available chlorine, 4.00-4.99%
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
: HENKE SASS WOLF
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Universal Filter Tips
: VWR Collection
VWR®, Bottle Top Dispensers
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection