Order Entry
Italy
ContactUsLinkComponent
 
 
Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™
  06220-1KG
 :  
 
Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™
  06220-1KG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 10 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 10 ppm
  • Sodium (Na):
    Max. 10 ppm
  • Nickel (Ni):
    Max. 10 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Vapor Pressure:
    1 mmHg ( 100 °C)

 

 

 :
 :133.34 g/mol
 :13,00335
 :190 °C (2.500 hPa)
 :2.44 g/cm³ (20 °C)
 :Ambient
 :231-208-1
 :1726
 :8,II
 :GHS05
 :7446-70-0
 :AlCl₃
 :MFCD00003422