Calcium chloride, anhydrous ≥97%, powder, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1.900 °C (1.013 hPa)
Melting Pt:765 °C
Density:2.150 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Colour of the solution:complying
- Clearness of the solution:complying
- Assay:Min. 97.0 %
- Mg and alkali salts:Max. 3.0 %
Specification Test Results
Frequently Bought Together