Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
  95682-2.5L
 :  
undefined
 
 
 
p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
  95682-2.5L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Refractive index (n 20/D):
    1.494 - 1.496
  • Non-volatile matter:
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • o-Xylene (GC):
    Max. 0.5 %
  • m-Xylole (GC):
    Max. 0.5 %
  • Sum of the isomers (GC):
    Max. 0.5 %
  • Benzene (GC):
    Max. 1.0 %
  • Ethyl benzene (GC):
    Max. 1.0 %
  • Toluene (GC):
    Max. 0.5 %
  • Refractive Index:
    n20/D 1.495

 

 

 :
 :106.17 g/mol
 :12,10214
 :137-139 °C (1.013 hPa)
 :137…139 °C (1.013 hPa)
 :12 °C
 :0.86 g/cm³ (20 °C)
 :25 °C
 :203-396-5
 :1307
 :3,III
 :GHS02
 :GHS08
 :GHS07
 :Ambient
 :106-42-3
 :C₆H₄(CH₃)₂
 :MFCD00008556