Order Entry
Italy
ContactUsLinkComponent
Sodium dihydrogen phosphate dihydrate 98-100.5% (dried basis), meets analytical specification of BP, Ph. Eur., USP, FCC, E339, Fluka™
Sodium dihydrogen phosphate dihydrate 98-100.5% (dried basis), meets analytical specification of BP, Ph. Eur., USP, FCC, E339, Fluka™
Nome del fornitore:  Honeywell Chemicals
Certificati
Sodium dihydrogen phosphate dihydrate 98-100.5% (dried basis), meets analytical specification of BP, Ph. Eur., USP, FCC, E339, Fluka™
Nome del fornitore:  Honeywell Chemicals
Certificati

Risultati dei test sulle specifiche

  • Assay (calculated to the dried substance):
    98.0 - 100.5 %
  • Assay (calculated on the anhydrous basis):
    98.0 - 103.0 %
  • Loss on drying (130°C):
    21.5 - 24.0 %
  • Water (Karl Fischer):
    18.0 - 26.5 %
  • Water insoluble matter:
    Max. 0.2 %
  • pH (5 %, 25 °C):
    4.2 - 4.5
  • pH (1 %,20°C):
    4.1 - 5.0
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 0.002 %
  • Cadmium (Cd):
    Max. 1 ppm
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 5 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Magnesium (Mg):
    Max. 0.002 %
  • Lead (Pb):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Al, Ca and Related Substances:
    complying
  • Chloride (Cl):
    Max. 50 ppm
  • Fluoride (F):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 200 ppm
  • Reducing impurities:
    complying
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying
  • Appearance:
    complying

Risultati dei test sulle specifiche

Informazioni correlate

Frequently Bought Together