Order Entry
Italy
ContactUsLinkComponent
Ethylene carbonate for synthesis
Corrosive
Ethylene carbonate for synthesis
Catalog # 8.44011.1000
Supplier:  Merck
CAS Number:  
Corrosive
Ethylene carbonate for synthesis
Catalog # 8.44011.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    PE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :88.06 g/mol
Boiling Pt:248 °C (1013 hPa)
Density:1.32 g/cm³ (40 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:143 °C
 :GHS05
CAS Number:96-49-1
Formula:C₃H₄O₃
MDL Number:MFCD00005382