Supelco® TraceCERT® Organic Reference Standard, Toluene-d₈ solution
CRM48593
: Merck
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Concentration:2000 μg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- CAS No.:2037-26-5
- Description:Toluene-d₈ solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Flash point:9,7 °C - Closed cup
- GHS signal: GHS02, GHS06, GHS08
: Certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311