HPLC columns, PAH analysis, NUCLEODUR® C18 PAH
Supplier: MACHEREY-NAGEL
Certificates
About this item
Special octadecyl modified silica phase for HPLC analysis of PAHs.
- High purity spherical silica gel
- Polymeric octadecyl modification
- Allows efficient gradient separation of PAHs according to EPA and EFSA
NUCLEODUR® C18 PAH columns are recommended for a gradient separation of 16 PAHs in accordance with EPA. An ultra-fast separation of 16 EPA PAHs is possible with 1,8 µm UHPLC phase. Moreover, separations of further PAH compounds such as PAHs according to EFSA can also be realised.
Selectivity: matched for PAH analysis.
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Specifications
- Catalog No:
- MANA760786.40
- MANA760773.30
- MANA761971.30
- MANA760784.40
- MANA761970.30
- MANA760784.30
- MANA760785.30
- MANA760785.40
- MANA760773.20
- MANA760786.30
- MANA760773.40
- MANA761970.20
- MANA760783.30
- MANA760783.40
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1