Order Entry
Italy
ContactUsLinkComponent
Analytical standard, Flonicamid metabolite TFNG, reference material, Honeywell Fluka™
Analytical standard, Flonicamid metabolite TFNG, reference material, Honeywell Fluka™
Catalog # HONCR1282-25MG
CAS Number:  
Analytical standard, Flonicamid metabolite TFNG, reference material, Honeywell Fluka™
Catalog # HONCR1282-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    207502-65-6
  • Description:
    Flonicamid metabolite TFNG
  • Melting point:
    153…155 °C
  • Formula:
    C₉H₇F₃N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    Flonicamid metabolite TFNG

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results