Analytical standard, Trifloxystrobin Metabolite CGA 321113, reference material, Honeywell Fluka™
Catalog # HONCR1275-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:252913-85-2
- Description:Trifloxystrobin Metabolite CGA 321113
- Melting point:114…116 °C
- Formula:C₁₉H₁₇F₃N₂O₄
- Storage temperature:Ambient
- Description:Trifloxystrobin Metabolite CGA 321113
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results