Order Entry
Italy
ContactUsLinkComponent
Analytical standard, Cloransulam-methyl, reference material, Honeywell Fluka™
Analytical standard, Cloransulam-methyl, reference material, Honeywell Fluka™
Catalog # HONCR1258-50MG
CAS Number:  
Analytical standard, Cloransulam-methyl, reference material, Honeywell Fluka™
Catalog # HONCR1258-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    147150-35-4
  • Description:
    Cloransulam-methyl
  • Melting point:
    214…219 °C
  • Formula:
    C₁₅H₁₃ClFN₅O₅S
  • Storage temperature:
    Ambient
  • Description:
    Cloransulam-methyl

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results