Order Entry
Italy
ContactUsLinkComponent
 
Analytical standard, 2,5-Dimethyl-celecoxib, reference material, Honeywell Fluka™
  HONCR1237-10MG
 :  
Analytical standard, 2,5-Dimethyl-celecoxib, reference material, Honeywell Fluka™
  HONCR1237-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    457639-26-8
  • Description:
    2,5-Dimethyl-celecoxib
  • Melting point:
    138…141 °C
  • Formula:
    C₁₈H₁₆F₃N₃O₂S
  • Storage temperature:
    Ambient
  • Description:
    2,5-Dimethyl-celecoxib

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results