Order Entry
Italy
ContactUsLinkComponent
Analytical standard, Fluthiacet-methyl, reference material, Honeywell Fluka™
Analytical standard, Fluthiacet-methyl, reference material, Honeywell Fluka™
  HONCR1195-25MG
 :  
Analytical standard, Fluthiacet-methyl, reference material, Honeywell Fluka™
  HONCR1195-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    117337-19-6
  • Description:
    Fluthiacet-methyl
  • Melting point:
    105…106.5 °C
  • Formula:
    C₁₅H₁₅ClFN₃O₃S₂
  • Storage temperature:
    Ambient
  • Description:
    Fluthiacet-methyl
  • Boiling point:
    249 °C

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results