Order Entry
Italy
ContactUsLinkComponent
Analytical standard, Pyraflufen-ethyl, reference material, Honeywell Fluka™
Analytical standard, Pyraflufen-ethyl, reference material, Honeywell Fluka™
Catalog # HONCR1082-10MG
CAS Number:  
Analytical standard, Pyraflufen-ethyl, reference material, Honeywell Fluka™
Catalog # HONCR1082-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    129630-19-9
  • Description:
    Pyraflufen-ethyl ≥98%
  • Melting point:
    125…130 °C
  • Formula:
    C₁₅H₁₃Cl₂F₃N₂O₄
  • Storage temperature:
    2…8 °C
  • Description:
    Pyraflufen-ethyl

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results