Cell viability assay kits, OZBlue
Supplier: OZ BIOSCIENCES
Certificates
About this item
OZBlue cell viability kit is a ready to use assay system based on the fluorimetric/colorimetric quantification of metabolic activity in living cells. Resazurin dye is used as an indicator of cell viability.
- Non toxic: Allows additional analysis on proliferating cells (mRNA, cytogenetic, apoptosis, immunophenotyping) and continuous monitoring of cultures over the time
- High sensitivity: More sensitive than (³H) thymidine incorporation and tetrazolium reduction assay (MTT, XTT)
- Simple protocol: No need for washing or extraction procedures
- Versatile: Can be used on cell lines, primary cells, bacteria, fungi
- Flexible: Colorimetric and fluorescence detection allows compatibility with flow cytometry, plate readers and microscopy detection
Store at +4 °C.
Frequently Bought Together