

Citric acid, anhydrous 99.5-100.05% (dry basis)
:77-92-9
:
:HOC(COOH)(CH₂COOH)₂
:MFCD00011669
:192.13 g/mol
:14,02326
:153…154 °C
:1.665
:201-069-1
:01-2119457026-42-0000
:GHS05
:GHS07
:Ambient
- Assay:99.5 - 100.5% (dry basis)
- Assay, Readily Carbonisable Substances:Passes Test
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Impurity Content, Oxalate:Passes Test
- Impurity Content, Sulphate (SO₄):Passes test
- Water Content (Karl Fischer Titration):≤1.0%
- Heavy Metals (Pb):≤10 µg/gm
Frequently Bought Together