Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TAK-242
TAK-242 ≥98% (by HPLC), Calbiochem®
TAK-242 ≥98% (by HPLC), Calbiochem®
TAK-242 ≥98% (by HPLC), Calbiochem®
614316-5MG
:
Merck Millipore (Calbiochem)
:
TAK-242 ≥98% (by HPLC), Calbiochem®
614316-5MG
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle
Pk:
5 mg
Seq:
2210
Purity:
≥98% by HPLC
:
:
361.8 g/mol
:
Packaged under inert gas.
:
GHS07
:
Refrigerator
:
243984-11-4
:
C₁₅H₁₇NO₄ClFS
:
MFCD11046362