
Geranyl Pyrophosphate triammonium salt ≥95% in methanol
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:116057-55-7
:
Formula:C₁₀H₂₀O₇P₂·3NH₃
MDL Number:MFCD00189726
:365.30 g/mol
:GHS02
Storage Temperature:Freezer
Specification Test Results
- Purity:≥95%
Specification Test Results
Frequently Bought Together