Order Entry
Italy
ContactUsLinkComponent
 
Corrosive
Hydrochloric acid 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., NF, USP volumetric solution
Catalog # 30024.980
Supplier:  VWR Collection
CAS Number:  
Corrosive
Hydrochloric acid 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., NF, USP volumetric solution
Catalog # 30024.980
Supplier:  VWR Collection
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    950 L
  • Seq:
    5783

Specifications

Specification Test Results

  • Conforms to Reag. Ph.Eur.:
    Passes test
  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • Conforms to USP/NF:
    Passes test
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
Density:1.02 g/cm³
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324